Isobutyronitrile 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Isopropyl cyanide

122530010
ACRO122530010EA 109 GBP
ACRO122530010
Isobutyronitrile 99%
Isobutyronitrile
Formula: C₄H₇N
MW: 69.11 g/mol
Boiling Pt: 107
Melting Pt: –72 °C
Density: 0.76 g/cm³
Flash Pt: 8
MDL Number: MFCD00001873
CAS Number: 78-82-0
EINECS: 201-147-5
UN: 2284
ADR: 3 II
Merck Index: 13,05173

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3720 to 1.3740 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR