Formaldehyde 37% w/w water Stabilised

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

Stabilisation: 5-15% MeOH

119690010 119690025 119690050 119690250
ACRO119690010EA 61.1 GBP
ACRO119690010 ACRO119690025 ACRO119690050 ACRO119690250
Formaldehyde 37% w/w water Stabilised
Formaldehyde
Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: -19.3
Melting Pt: –15 °C
Density: 1.083 g/cm³
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 1198
ADR: 3 III
Merck Index: 13,04259

Order Now

Specification Test Results

Appearance Clear solution
Color scale ≤10 APHA
Stabilizer 5 to 15 % methanol
Titration 35.0 to 39.0 %
Free acid (HCOOH) ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR