N,N-Dimethylaniline 99%

Supplier: Thermo Fisher Scientific
Danger

115920010 115920025 115920050 115920100
ACRO115920010EA 90 GBP
ACRO115920010 ACRO115920025 ACRO115920050 ACRO115920100
N,N-Dimethylaniline 99%
N,N-Dimethylaniline
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 193…194 °C (760 mmHg)
Melting Pt: 1.5…2.5 °C
Density: 0.95 g/cm³
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00008304
CAS Number: 121-69-7
EINECS: 204-493-5
UN: 2253
ADR: 6.1 II
Merck Index: 13,03261

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5570 to 1.5590 (20°C, 589 nm)
GC ≥98.5 %
Titration with HClO4 ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR