Citric acid, anhydrous 99%, pure

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

110450010 110450050 110450250
ACRO110450010EA 106 GBP
ACRO110450010 ACRO110450050 ACRO110450250
Citric acid, anhydrous 99%, pure
Citric acid
Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Boiling Pt: 310
Melting Pt: 153 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
REACH: 01-2119457026-42-0000
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystals
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %
GC ≥98.5 %
Specific optical rotation -43.2° to -45.2° (21°C, 589 nm) (c=3, 1,2-dichloroethane)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR