Avantor® ACE® C4-300, HPLC Columns, 5 µm
Supplier: Avantor
ACE-223-0246EA 0 GBP
ACE-223-0246 ACE-223-0502 ACE-223-0503 ACE-223-0510 ACE-223-0520 ACE-223-0546 ACE-223-1002 ACE-223-1003 ACE-223-1010 ACE-223-1020 ACE-223-1046 ACE-223-1220 ACE-223-1246 ACE-223-1502 ACE-223-1503 ACE-223-1510 ACE-223-1520 ACE-223-1546 ACE-223-2502 ACE-223-2503 ACE-223-2504 ACE-223-2510 ACE-223-2520 ACE-223-2546 ACE-223-7502 ACE-223-7520 ACE-223-0102GD ACE-223-0103GD ACE-223-0110GD ACE-223-7530 ACE-223-25003 ACE-223-25001 ACE-223-005003GD ACE-223-2500075
Avantor® ACE® C4-300, HPLC Columns, 5 µm
Chromatography Columns
Avantor® ACE® C4-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
  • Designed for peptide and protein separations
  • Method validation kits contain three different batches of the same phase - available on request