Avantor® ACE® C8-300, HPLC Columns, 3 µm
Supplier: Avantor
ACE-212-0501EA 0 GBP
ACE-212-0501 ACE-212-0502 ACE-212-0503 ACE-212-0504 ACE-212-0546 ACE-212-1002 ACE-212-1003 ACE-212-1004 ACE-212-1046 ACE-212-1502 ACE-212-1546 ACE-212-7502 ACE-212-0102GD ACE-212-0103GD ACE-212-10005S ACE2121546MVK
Avantor® ACE® C8-300, HPLC Columns, 3 µm
Chromatography Columns
Avantor® ACE® C8-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
  • Designed for peptide and protein separations
  • Method validation kits contain three different batches of the same phase - available on request