Anti-IL17RB Rabbit Polyclonal Antibody

Supplier: Abgent
AP6762C
ABGEAP6762CEA 345 GBP
ABGEAP6762C
Anti-IL17RB Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL17RB (interleukin 17 receptor B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR