Anti-PROSC Rabbit Polyclonal Antibody

Supplier: Abgent
AP13026B
ABGEAP13026BEA 345 GBP
ABGEAP13026B
Anti-PROSC Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PROSC (proline synthetase co-transcribed homolog (bacterial))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR