Anti-EIF4E Mouse Monoclonal Antibody

Supplier: Abgent
AM1852B
ABGEAM1852BEA 345 GBP
ABGEAM1852B
Anti-EIF4E Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: EIF4E (eukaryotic translation initiation factor 4E)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR