Anti-Major latex allergen Hev b 5 Mouse Monoclonal Antibody [clone: 5-11.6]

Supplier: Abcam
AB138707-100
ABCAAB138707-100EA 1080 GBP
ABCAAB138707-100
Anti-Major latex allergen Hev b 5 Mouse Monoclonal Antibody [clone: 5-11.6]
Antibodies
Type: Primary
Antigen: Major latex allergen Hev b 5
Clonality: monoclonal
Clone: 5-11.6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR