SYBR Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Supplier: AAT BIOQUEST
67008
AATB67008EA 120 GBP
AATB67008
SYBR Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Microplates PCR Plates
The SYBR dye qPCR calibration plate can be used to maintain the 7500 real-time PCR system with fast 96-well block.
  • The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
  • This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.
Caution: Avoid freeze-thawing plates. It is not recommended to use the plates moreMore Product Information