Transfectamine™ 5000 Transfection Reagent
Supplier: AAT BIOQUEST
60020 60021 60022
AATB60020EA 271 GBP
AATB60020 AATB60021 AATB60022
Transfectamine™ 5000 Transfection Reagent
Transfection Reagents
Transfectamine™ 5000 Transfection Reagent is a powerful and versatile transfection reagent for the introduction of nucleic acids into eukaryotic cells, or more specifically, into animal cells.
  • Easy-to-use, minimal steps required
  • High transfection efficiency
  • Works with a wide variety of eukaryotic cells
  • Effective on dissociated cells and slices
Caution: For research use only.