Dextran, M.W. 10,000 Cal-520™ labelled

Supplier: AAT BIOQUEST

20601
AATB20601EA 406 GBP
AATB20601
Dextran, M.W. 10,000 Cal-520™ labelled
Dextran

Calcium measurement is critical for numerous biological investigations.


  • Well-retained in cells
  • 10000 mw
  • Useful for in vivo imaging


From Eukaryotic cells


Caution: For research use only.

Formula: (C₆H₁₀O₅)n
MDL Number: MFCD00130935
CAS Number: 9004-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR