Gelite™ Orange Nucleic Acid Gel Staining Kit
Supplier: AAT BIOQUEST
17594
AATB17594EA 271 GBP
AATB17594
Gelite™ Orange Nucleic Acid Gel Staining Kit
Assays Nucleic Acid Assays
Gelite™ Orange is an extremely sensitive nucleic acid gel stain for detecting DNA or RNA in gels using a standard 300 nm UV transilluminator and Polaroid 667 black-and-white print film.
  • High sensitivity
  • Exceptional DNA affinity
  • Robust protocol
  • Multiple applications
Caution: For research use only.