3'-O-Azidomethyl-dATP
Supplier: AAT BIOQUEST
17220 17221
AATB17220EA 594 GBP
AATB17220 AATB17221
3'-O-Azidomethyl-dATP
Nucleic Acid Reagents Nucleotides
3'-O-Azidomethyl-dATP is one of the four essential dNTPs (A, C, G, T) that can be used for the DNA sequencing by synthesis (SBS).
  • Highly purified
DNA sequencing is a fundamental tool in biological and medical research (especially for personalised medicine).
Caution: For research use only.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more