iFluor® 594 fluorescent probe

Supplier: AAT BIOQUEST
Warning

1018
AATB1018EA 258 GBP
AATB1018
iFluor® 594 fluorescent probe
iFluor®

The tetrazine-trans-cyclooctene (TCO) ligation constitutes a non-toxic biomolecule labeling method of unparalleled speed.


  • pH-insensitive
  • Azide reactive
  • High photostability
  • Brighter signal

ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c6eae9d1873cc8ee60ead446bef88aa326aff1c&vid=07b2fb15cac2bd20b9968271ed609b2da1ded5a0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR