Phenazone, Supelco®

Supplier: Merck
Warning

Synonyms: 2,3-Dimethyl-1-phenyl-3-pyrazolin-5-one, Antipyrine

A5882-25G A5882-500G A5882-100G
A5882-25GEA 19.9 GBP
A5882-25G A5882-500G A5882-100G
Phenazone, Supelco®
Phenazone
Formula: C₁₁H₁₂N₂O
MW: 188.23 g/mol
Boiling Pt: 388
Melting Pt: 152…154 °C
Density: 1.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003146
CAS Number: 60-80-0
EINECS: 200-486-6
Merck Index: 12,00757
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bec02f544f0936850e3e272ec84ae82307c114e&vid=3bec02f544f0936850e3e272ec84ae82307c114e&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee4dd66725fcf5865f4f2fd94c21c72f5de2192f&vid=ee4dd66725fcf5865f4f2fd94c21c72f5de2192f&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0dd4ec2287655652829af0f426e804a714c0756a&vid=0dd4ec2287655652829af0f426e804a714c0756a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR