Formamide 99,5 N_ASSAY_METHOD deionized Molecular_Biology

Supplier: PanReac AppliChem
Danger

Synonyms: Carbamaldehyde

A2156.0500 A2156.1000
A2156.0500EA 115 GBP
A2156.0500 A2156.1000
Formamide 99,5 N_ASSAY_METHOD deionized Molecular_Biology
Formamide

  • n20/D 1.4472


Caution: For scientific research and laboratory use only.

Formula: HCONH₂
MW: 45.04 g/mol
Boiling Pt: 210
Melting Pt: 2
Density: 1.1334 g/cm³ (20 °C)
Flash Pt: 79
Storage Temperature: Ambient
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

DNases/RNases/Proteases not detectable
Assay (from N) min. 99.5 %
Water max. 0.1 %
Chloride max. 0.00005 %
Fe max. 0.00001 %
PB max. 0.00001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR