2,5-Dimethoxybenzaldehyde 98

Supplier: Thermo Fisher Scientific
Warning

A19928.14 A19928.22 A19928.36
A19928.14EA 26.9 GBP
A19928.14 A19928.22 A19928.36
2,5-Dimethoxybenzaldehyde 98
2,5-Dimethoxybenzaldehyde

Purity: 98+% CAS No.: 93-02-7 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Melting Point: 47-51° Boiling Point: 146°/10mm Beilstein Reference: 509301 MDL No.: MFCD00003314

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 146 °C (10 mmHg)
Melting Pt: 47…51 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003314
CAS Number: 93-02-7
EINECS: 202-211-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR