Dapsone 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone

A18976.22 A18976.36
A18976.22EA 39.9 GBP
A18976.22 A18976.36
Dapsone 98%
Dapsone

Purity: 98% CAS No.: 80-08-0 Molecular Formula: C12H12N2O2S Formula Weight: 248.30 Odour: Odourless Melting Point: 175-178° Beilstein Reference: 788055 Merck Reference: 14,2822 MDL No.: MFCD00007887

Formula: C₁₂H₁₂N₂O₂S
MW: 248.31 g/mol
Melting Pt: 175…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00007887
CAS Number: 80-08-0
EINECS: 201-248-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR