Butyryl chloride ≥98%

Supplier: Thermo Fisher Scientific
Danger

A17575.30 A17575.0B A17575.18
A17575.30EA 28 GBP
A17575.30 A17575.0B A17575.18
Butyryl chloride ≥98%
Butyryl chloride

Purity: 98% CAS No.: 141-75-3 Molecular Formula: CH3CH2CH2COCl Formula Weight: 106.55 Flash Point: 18°(64°F) Refractive Index: 1.4120 Odour: Acrid Melting Point: -89° Boiling Point: 101-102° Beilstein Reference: 605395 Merck Reference: 14,1598 UN No.: 2353 MDL No.: MFCD00000752

Formula: C₄H₇ClO
MW: 106.55 g/mol
Boiling Pt: 101…102 °C
Melting Pt: –89 °C
Density: 1.026
Flash Pt: 18 °C (64 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000752
CAS Number: 141-75-3
EINECS: 205-498-5
UN: 2353
ADR: 3 II
Merck Index: 13,01598

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR