Murexide

Supplier: Thermo Fisher Scientific

Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt

A17540.09 A17540.18 A17540.30
A17540.09EA 41.4 GBP
A17540.09 A17540.18 A17540.30
Murexide
Murexide

CAS No.: 3051-09-0 Molecular Formula: C8H8N6O6 Formula Weight: 284.19 Melting Point: 300° Beilstein Reference: 3582175 Merck Reference: 14,6306 MDL No.: MFCD00012777

Formula: C₈H₈N₆O₆
MW: 284.19 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00012777
CAS Number: 3051-09-0
EINECS: 221-266-6
Merck Index: 13,06330

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR