Hexamethylenetetramine 99

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Hexamethylenetetramine, Hexamine, Urotropine, Formin

A17213.30 A17213.0B A17213.0I
A17213.30EA 22 GBP
A17213.30 A17213.0B A17213.0I
Hexamethylenetetramine 99
Methenamine

Purity: 99+% CAS No.: 100-97-0 Molecular Formula: C6H12N4 Formula Weight: 140.19 Flash Point: 250°(482°F) Melting Point: ca 280° subl. Beilstein Reference: 2018 Merck Reference: 14,5966 UN No.: 1328 MDL No.: MFCD00006895

Formula: C₆H₁₂N₄
MW: 140.19 g/mol
Melting Pt: ∼ 280 °C (subl.)
Density: 1.33
Flash Pt: 250 °C (482 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006895
CAS Number: 100-97-0
EINECS: 202-905-8
UN: 1328
ADR: 4.1 III
Merck Index: 13,05994
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3ecbb5df5394322c2150525ce6e9f48b5aadde7&vid=04b623f7f333c7cf159016619cf91fc792af9a3f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR