(4-tert-Butylphenoxy)acetic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-(4-(tert-Butyl)phenoxy)acetic acid

A17017.06 A17017.14 A17017.22
A17017.06EA 15.7 GBP
A17017.06 A17017.14 A17017.22
(4-tert-Butylphenoxy)acetic acid ≥98%
(4-tert-Butylphenoxy)acetic acid

Purity: 98% CAS No.: 1798-04-5 Molecular Formula: C12H16O3 Formula Weight: 208.26 MDL No.: MFCD00021758

Formula: (C₆H₄(C(CH₃)₃))CH₂COOH
MW: 208.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00021758
CAS Number: 1798-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR