L(+)-Aspartic acid

Supplier: PanReac AppliChem

Synonyms: (S)-(+)-Aminosuccinic acid (S)-Aminobutanedioic acid (S)-Aminosuccinic acid H-Asp-OH L-Asparaginic acid L(+)-Aspartic acid, (S)-(+)-Aspartic acid, L(+)-2-Amino-1,4-butanedioic acid, (S)-(+)-2-Amino-1,4-butanedioic acid, L-Aspartic acid, H-Asp-OH, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, (S)-2-Aminosuccinic acid, Asp, D

A1701.1000
A1701.1000EA 180 GBP
A1701.1000
L(+)-Aspartic acid
L(+)-Aspartic acid
Formula: C₄H₇NO₄
MW: 133.1 g/mol
Boiling Pt: 311
Melting Pt: 251
Density: 1.6613 g/cm³ (13 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002616
CAS Number: 56-84-8
EINECS: 200-291-6
Merck Index: 13,00845

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR