Canada balsam, filtered_natural

Supplier: Thermo Fisher Scientific
Warning

A16289.14 A16289.22 A16289.36
A16289.14EA 31.9 GBP
A16289.14 A16289.22 A16289.36
Canada balsam, filtered_natural
Canada balsam

CAS No.: 8007-47-4 Flash Point: 43°(109°F) Refractive Index: 1.5230 Merck Reference: 14,946 UN No.: 1993 MDL No.: MFCD00132800


Mounting medium for microscopy

Density: 0.99
Flash Pt: 43 °C (109 °F)
Storage Temperature: Ambient
MDL Number: MFCD00132800
CAS Number: 8007-47-4
EINECS: 232-362-2
UN: 1993
ADR: 3 III
Merck Index: 14,946

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR