Nickel(II) nitrate hexahydrate ≥98%

Supplier: Thermo Fisher Scientific
Danger

A15540.0B A15540.0I A15540.22 A15540.36 A15540.A4
A15540.0BEA 90 GBP
A15540.0B A15540.0I A15540.22 A15540.36 A15540.A4
Nickel(II) nitrate hexahydrate ≥98%
Nickel (II) nitrate hexahydrate

Purity: 98% CAS No.: 13478-00-7 Molecular Formula: N2NiO6.6H2O Formula Weight: 290.80 (182.70anhy) Odour: Odourless Melting Point: 56.7° Boiling Point: 137° Merck Reference: 14,6513 UN No.: 2725 MDL No.: MFCD00149805

Formula: Ni(NO₃)₂·6H₂O
MW: 290.79 g/mol
Boiling Pt: 137 °C
Melting Pt: 56.7
Density: 2.05
Storage Temperature: Ambient
MDL Number: MFCD00149805
CAS Number: 13478-00-7
EINECS: 236-068-5
UN: 2725
Merck Index: 12,06596

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR