4-Aminoantipyrine 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone, 4-Aminophenazone

A13846.0E A13846.22 A13846.36
A13846.0EEA 953 GBP
A13846.0E A13846.22 A13846.36
4-Aminoantipyrine 97%
4-Aminoantipyrine

Purity: 97% CAS No.: 83-07-8 Molecular Formula: C11H13N3O Formula Weight: 203.25 Physical Form: Powder Appearance: Yellow to pale brown Melting Point: 106-110° Beilstein Reference: 181635 Merck Reference: 14,591 MDL No.: MFCD00003145

Formula: C₁₁H₁₃N₃O
MW: 203.24 g/mol
Melting Pt: 106…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003145
CAS Number: 83-07-8
EINECS: 201-452-3
Merck Index: 13,00596

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR