Diethyl phenylthiomethylphosphonate 96%

Supplier: Thermo Fisher Scientific

A13660.09 A13660.18
A13660.09EA 75.9 GBP
A13660.09 A13660.18
Diethyl phenylthiomethylphosphonate 96%
Diethyl phenylthiomethylphosphonate

Purity: 96% CAS No.: 38066-16-9 Molecular Formula: C11H17O3PS Formula Weight: 260.29 Refractive Index: 1.5340 Boiling Point: 146-148°/0.5mm Beilstein Reference: 1246215 MDL No.: MFCD00015130

Formula: C₁₁H₁₇O₃PS
MW: 260.288 g/mol
Boiling Pt: 146…148 °C (0.5 mmHg)
Density: 1.174
Storage Temperature: Ambient
MDL Number: MFCD00015130
CAS Number: 38066-16-9
EINECS: 253-770-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR