DL-Methionine 99

Supplier: Thermo Fisher Scientific

Synonyms: 2-Amino-4-(methylthio)butanoic acid, (±)-2-Amino-4-(methylthio)butyric acid, Methioninum racemicum, Methionine racematic, H-DL-Met-OH, (±)-Methionine, (±)-2-Amino-4-(methylthio)butanoic acid

A11457.0B A11457.0I A11457.30
A11457.0BEA 75.2 GBP
A11457.0B A11457.0I A11457.30
DL-Methionine 99
DL-Methionine

Purity: 99% CAS No.: 59-51-8 Molecular Formula: C5H11NO2S Formula Weight: 149.21 Melting Point: ca 272° dec. Beilstein Reference: 636185 Merck Reference: 14,5975 MDL No.: MFCD00063096

Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
Melting Pt: ∼ 272 °C (dec.)
Density: 1.34
Storage Temperature: Ambient
MDL Number: MFCD00063096
CAS Number: 59-51-8
EINECS: 200-432-1
Merck Index: 13,06004

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR