Paraformaldehyde 97

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Polyoxymethylene

A11313.0C A11313.0E A11313.22 A11313.36 A11313.PG
A11313.0CEA 239 GBP
A11313.0C A11313.0E A11313.22 A11313.36 A11313.PG
Paraformaldehyde 97
Paraformaldehyde
Formula: (CH₂O)n
Boiling Pt: 135…136 °C (13 mmHg)
Melting Pt: 120…170 °C (dec.)
Density: 1.30
Flash Pt: 71 °C (159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00133991
CAS Number: 30525-89-4
EINECS: 200-001-8
UN: 2213
ADR: 4.1 III
Merck Index: 14,7025

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR