Barbituric acid 99

Supplier: Thermo Fisher Scientific

Synonyms: N,N'-Malonylurea, 2,4,6-Trihydroxypyrimidine

A10934.14 A10934.22 A10934.36
A10934.14EA 23.5 GBP
A10934.14 A10934.22 A10934.36
Barbituric acid 99
Barbituric acid

Purity: 99% CAS No.: 67-52-7 Molecular Formula: C4H4N2O3 Formula Weight: 128.09 Flash Point: 150°(302°F) pH: 2-3 Melting Point: ca 250° dec. Beilstein Reference: 120502 Merck Reference: 14,963 MDL No.: MFCD00006666

Formula: C₄H₄N₂O₃
MW: 128.09 g/mol
Boiling Pt: 441
Melting Pt: ∼ 250 °C (dec.)
Flash Pt: 150 °C (302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006666
CAS Number: 67-52-7
EINECS: 200-658-0
Merck Index: 13,00966

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR