EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) 98+%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine monohydrochloride, N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide monohydrochloride, 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDC hydrochloride, EDC, EDAC HCl, EDAC hydrochloride, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, WSC HCl, EDC HCl

A10807.14 A10807.06 A10807.03
A10807.14EA 275 GBP
A10807.14 A10807.06 A10807.03
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) 98+%
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride

Purity: 98+% CAS No.: 25952-53-8 Molecular Formula: C8H17N3.HCl Formula Weight: 191.70 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Moisture Sensitive Appearance: White Melting Point: ca 113° Beilstein Reference: 5764110 MDL No.: MFCD00012503

Formula: C₈H₁₇N₃·HCl
MW: 191.7 g/mol
Melting Pt: ∼ 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012503
CAS Number: 25952-53-8
EINECS: 247-361-2
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d8bf2864bcf21ffe6dcca6f22388d1fe427e22&vid=9080dd6b66a1622ef32a2da2138f85e44cc881b8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR