Isoniazid 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Picolinic acid hydrazide, 4-Pyridinecarboxylic acid hydrazide, Isonicotinic hydrazide, Pyridine-4-carboxylic acid hydrazide, Isonicotinic acid hydrazide

A10583.0B A10583.18 A10583.22 A10583.36
A10583.0BEA 195 GBP
A10583.0B A10583.18 A10583.22 A10583.36
Isoniazid 98
Isoniazid

Purity: 98+% CAS No.: 54-85-3 Molecular Formula: C6H7N3O Formula Weight: 137.14 Melting Point: 169-174° Beilstein Reference: 119374 Merck Reference: 14,5186 MDL No.: MFCD00006426

Formula: C₆H₇N₃O
MW: 137.14 g/mol
Melting Pt: 169…174 °C
Density: 1.42 g/cm³ (20 °C)
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00006426
CAS Number: 54-85-3
EINECS: 200-214-6
Merck Index: 13,05205

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR