tert-Butylacetyl chloride ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3,3-Dimethylbutyryl chloride

A10162.06 A10162.14 A10162.22 A10162.36
A10162.06EA 19 GBP
A10162.06 A10162.14 A10162.22 A10162.36
tert-Butylacetyl chloride ≥98%
tert-Butylacetyl chloride

Purity: 98+% CAS No.: 7065-46-5 Molecular Formula: (CH3)3CCH2COCl Formula Weight: 134.61 Storage Temperature: 2°C to 8°C Flash Point: 21°(69°F) Refractive Index: 1.4230 Odour: Acrid Boiling Point: 128-130° Beilstein Reference: 1740421 UN No.: 2924 MDL No.: MFCD00000737

Formula: C₆H₁₁ClO
MW: 134.61 g/mol
Boiling Pt: 128…130 °C
Density: 0.967
Flash Pt: 21 °C (69 °F)
MDL Number: MFCD00000737
CAS Number: 7065-46-5
EINECS: 230-356-4
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR