4-Ethoxymethylene-2-phenyl-2-oxazoline-5-one 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (4E)-4-(Ethoxymethylene)-2-phenyl-1,3-oxazol-5(4H)-one, 4-Ethoxymethylene-2-phenyloxazolin-5-one

A10155.03 A10155.06
A10155.03EA 34 GBP
A10155.03 A10155.06
4-Ethoxymethylene-2-phenyl-2-oxazoline-5-one 98
4-Ethoxymethylene-2-phenyl-2-oxazoline-5-one

Purity: 98+% CAS No.: 15646-46-5 Molecular Formula: C12H11NO3 Formula Weight: 217.22 Storage Temperature: 2°C to 8°C Melting Point: 93-97° Beilstein Reference: 163055 MDL No.: MFCD00003204

Formula: C₁₂H₁₁NO₃
MW: 217.22 g/mol
Melting Pt: 93…97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03225480
CAS Number: 15646-46-5
EINECS: 239-713-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR