Milk, non fat (skimmed milk), powder

Supplier: PanReac AppliChem

A0830.0500 A0830.1000 A0830.5000
A0830.0500EA 49.4 GBP
A0830.0500 A0830.1000 A0830.5000
Milk, non fat (skimmed milk), powder
Non-fat milk powder (skim milk)
Storage Temperature: Ambient
CAS Number: 999999-99-4
EINECS: 310-127-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR