Propionic acid, Supelco®

Supplier: Merck
Danger

Synonyms: Propanoic acid

MQ100

94425-1ML-F 94425-5ML-F
94425-1ML-FEA 37.5 GBP
94425-1ML-F 94425-5ML-F
Propionic acid, Supelco®
Propionic acid
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 141
Melting Pt: –21 °C
Density: 0.99 g/cm³ (20 °C)
Flash Pt: 49
Storage Temperature: Ambient
MDL Number: MFCD00002756
CAS Number: 79-09-4
EINECS: 201-176-3
UN: 3463
ADR: 8 II
REACH: 01-2119486971-24
Merck Index: 13,07917
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d822519a8a4619297077959a9eb21c4b3a7a74f&vid=65e85f76282d4378293f82c345524df2403c3540&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b49923b3bc5ad7a51933ce096ddee00a8b6392d4&vid=daed86095bfff04a1b9f59ed786481b44a66e10e&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR