Dichloromethane, low water content ≥99.5% stabilised, BakerDRY™, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Methylene chloride

Stabilisation: Stabilised with cyclohexene

9295-12EA 103 GBP
9295-12
Dichloromethane, low water content ≥99.5% stabilised, BakerDRY™, J.T.Baker®
Dichloromethane

Specifically manufactured for use in organic, organometallic, and oligonucleotide synthesis requiring low water solvents.

Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.5 °C
Melting Pt: –95 °C
Density: 1.318 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use = 99.5 %
ACS - Assay (by GC, corrected for water, exclusive of preservative) ≥99.5 %
Appearance Passes Test
ACS - Color (APHA) ≤10
ACS - Free Halogens Passes Test
ACS - Preservative (cyclohexene), For Information Only = 2 ppm
ACS - Residue after Evaporation ≤2 ppm
ACS - Titrable Acid (µeq/g) ≤0.2
ACS - Water (by KF, coulometric) ≤60 ppm
Product Information (not specifications):
Boiling Point (typical)................. 39.5 °C.
Density (g/mL) at 25 °C (typical)........1.318


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR