1,4-Dioxane 99,0 GC, corrected for water content Stabilised, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

Stabilisation: 0.075% BHT

9231-22EA 331 GBP
9231-22 9231-03 9231-01 9231-R
1,4-Dioxane 99,0 GC, corrected for water content Stabilised, BAKER ANALYZED® ACS, J.T.Baker®
1,4-Dioxane

Caution: STORAGE: Store above 11°C (53°F) or product will freeze.

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 101.1
Melting Pt: 11.8
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 12 (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3 II
Merck Index: 13,03330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use = 99.0 %
Assay (C₄H₈O₂) (by GC, corrected for water) ≥99.0 %
Carbonyl Compounds (as HCHO) (by polarography) ≤0.01 %
Color (APHA) ≤20
Freezing Point ≥11.0 °C
Peroxide (as H₂O₂) ≤0.005 %
Residue after Evaporation ≤0.0050 %
Titrable Acid (meq/g) ≤0.0016
Water (by KF, coulometric) ≤0.05 %
STORAGE: Store above 11°C (53°F) or product will freeze.
Contains 0.075% Butylated Hydroxytoluene as a stabilizer.
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR