Supelco® Organic Reference Standard, Schisandrin A

Supplier: Merck

90139-50MG
90139-50MGEA 307 GBP
90139-50MG
Supelco® Organic Reference Standard, Schisandrin A
Standards Organic Standards
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=202b3c3fc29b39359d78d787a31b4a69d0d0a447&vid=202b3c3fc29b39359d78d787a31b4a69d0d0a447&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Synonym(s) Deoxyschisandrin; Deoxyschizandrin; Schizandrin A
Environmentally Preferable
M-Clarity quality level MQ100
CAS No. 61281-38-7
MDL No. MFCD09026934
Molecular weight 416,51
Shipping conditions STD
Storage conditions 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR