Magnesium standard for IC, TraceCERT®, Supelco®
Supplier: Merck
89441-100ML
89441-100MLEA 68.1 GBP
89441-100ML
Magnesium standard for IC, TraceCERT®, Supelco®
Standards Cationic Standards Magnesium Standards
This standard is prepared with high purity Mg(NO₃)₂, HNO₃ and water.
Certifications: Certified in accordance with ISO/IEC 17025 and ISO 17034