Petroleum spirit, 50…70 °C 90, AnalaR NORMAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

88263.290EA 70.8 GBP
88263.290 88263.360 88263.460
Petroleum spirit, 50…70 °C 90, AnalaR NORMAPUR®
Petroleum spirit
Density: 0.7
Flash Pt: -10
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 1268
ADR: 3 II

Order Now

Specification Test Results

Readily carbonisable substances passes test
Density (20/4) 0.654 - 0.670
Free acidity max. 0.00008 meq/g
Boiling range (50-70°C) (Vol%) min. 90%
Evaporation residue max. 10 ppm
Water max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR