Silver standard solution, 1000 mg/L Ag in 5% nitric acid (from Ag), standard for AAS, Specpure®

Supplier: Thermo Fisher Scientific

88096.AEEA 35.3 GBP
88096.AE
Silver standard solution, 1000 mg/L Ag in 5% nitric acid (from Ag), standard for AAS, Specpure®
Standards Cationic Standards Silver Standards

Molecular Formula: Ag in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00003397


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR