Sodium hydroxide 0.005 mol/l (0.005 N) methanol_50 Reag. Ph. Eur. 1081405

Supplier: VWR Chemicals
Danger

Synonyms: Caustic soda

87937.180EA 73.8 GBP
87937.180
Sodium hydroxide 0.005 mol/l (0.005 N) methanol_50 Reag. Ph. Eur. 1081405
Sodium hydroxide

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 3286
ADR: 3 (6.1, 8) II
Merck Index: 14,08627

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR