Zinc chloride, anhydrous 99,95 Metals basis

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride

87900.14 87900.22 87900.88
87900.14EA 69.1 GBP
87900.14 87900.22 87900.88
Zinc chloride, anhydrous 99,95 Metals basis
Zinc chloride

Purity: 99.95% CAS No.: 7646-85-7 Molecular Formula: Cl2Zn Formula Weight: 136.29 Physical Form: Lump Density: 2.91 Appearance: White Odour: Odourless Melting Point: 290° Boiling Point: 732° Merck Reference: 14,10132 UN No.: 2331 MDL No.: MFCD00011295

Formula: ZnCl₂
MW: 136.3 g/mol
Boiling Pt: 732 °C
Melting Pt: 290 °C
Density: 2.91
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8 III
Merck Index: 14,10132

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR