Orthophosphoric acid 10% Reag. Ph. Eur. 1065101

Supplier: VWR Chemicals
Warning

Synonyms: Phosphoric acid

87895.180EA 73.8 GBP
87895.180
Orthophosphoric acid 10% Reag. Ph. Eur. 1065101
Orthophosphoric acid

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: H₃PO₄
MW: 98 g/mol
Boiling Pt: 158
Melting Pt: 21
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8 III
Merck Index: 14,07344

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR