Nitrochromic reagent Reag. Ph. Eur. 1059100

Supplier: VWR Chemicals
Danger

87881.180EA 73.8 GBP
87881.180
Nitrochromic reagent Reag. Ph. Eur. 1059100
Nitrochromic reagent

The solution bottle should be open for the minimum time required to dispense the solution.
After use, the bottle should be tightly recapped and stored under normal laboratory conditions.


Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Storage Temperature: Ambient
MDL Number: MFCD00011367
UN: 3289
ADR: 6.1 (8) I

Order Now

Specification Test Results

Starting materials potassium dichromate R / nitric acid R
Performance test passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR