Hydrogen chloride 3 mol/l 1-butanol, Supelco®

Supplier: Merck


MQ100

87472-250ML-F 87472-50ML-F
87472-250ML-FEA 167 GBP
87472-250ML-F 87472-50ML-F
Hydrogen chloride 3 mol/l 1-butanol, Supelco®
Hydrogen chloride
Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45fdfff2ecf56445fd5588c533d60f8e9fd98ddf&vid=b9ca57b3071b503867a4396e65f0dafc5b76915e&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR