Methanol-[D₄] Isotope_(99.8% D) NMR

Supplier: VWR Chemicals
Danger

Synonyms: Methanol-[²H₄], Hydroxy methane-D4, Methanol-d4, Methyl-D3 alcohol-D, Carbinol-D4, Wood alcohol-D4, Methyl alcohol-D4, Tetradeuteromethanol

87157.0010EA 108 GBP
87157.0010 87157.0100 87157.0025 87157.0006 87157.0011
Methanol-[D₄] Isotope_(99.8% D) NMR
Methanol-[D4]
Formula: D₃COD
MW: 36.01 g/mol
Boiling Pt: 65.3 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3 (6.1) II

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.9 %
Isotopic enrichment (FT NMR 400 MHz)(D) Min. 99.80 %
Water (HDO+D2O) Max. 0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR